Anti-OR13G1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316472EA 619 CHF
ORIGTA316472
Anti-OR13G1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: OR13G1 (olfactory receptor, family 13, subfamily G, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR