Anti-OR51I2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316410EA 619 CHF
ORIGTA316410
Anti-OR51I2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: OR51I2 (olfactory receptor, family 51, subfamily I, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR