Anti-OR8D1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316300EA 642 CHF
ORIGTA316300
Anti-OR8D1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: OR8D1 (olfactory receptor, family 8, subfamily D, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR