Anti-ATP6V1C2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316265EA 642 CHF
ORIGTA316265
Anti-ATP6V1C2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: ATP6V1C2 (ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR