Anti-HIPK4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316152EA 619 CHF
ORIGTA316152
Anti-HIPK4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:00-1:500; ELISA: 1:20000

Type: Primary
Antigen: HIPK4 (homeodomain interacting protein kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR