Anti-BRI3BP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316127EA 656 CHF
ORIGTA316127
Anti-BRI3BP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: BRI3BP (BRI3 binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR