Anti-COX4I2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315934EA 656 CHF
ORIGTA315934
Anti-COX4I2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: COX4I2 (cytochrome c oxidase subunit IV isoform 2 (lung))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR