Anti-SLC52A2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315790EA 642 CHF
ORIGTA315790
Anti-SLC52A2 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: SLC52A2 (solute carrier family 52 (riboflavin transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR