Anti-ZNF446 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315595EA 656 CHF
ORIGTA315595
Anti-ZNF446 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ZNF446 (zinc finger protein 446)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR