Anti-PPP1R14D Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315541EA 642 CHF
ORIGTA315541
Anti-PPP1R14D Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: PPP1R14D (protein phosphatase 1, regulatory (inhibitor) subunit 14D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR