Anti-ANKRD26 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315188EA 642 CHF
ORIGTA315188
Anti-ANKRD26 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: ANKRD26 (ankyrin repeat domain 26)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR