Anti-MAP4K3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314756EA 656 CHF
ORIGTA314756
Anti-MAP4K3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: MAP4K3 (mitogen-activated protein kinase kinase kinase kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR