Anti-PPP1R3C Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314373EA 619 CHF
ORIGTA314373
Anti-PPP1R3C Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: PPP1R3C (protein phosphatase 1, regulatory subunit 3C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR