Anti-GRM6 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313975EA 642 CHF
ORIGTA313975
Anti-GRM6 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 500-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: GRM6 (glutamate receptor, metabotropic 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR