Anti-EPB41L2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313824EA 642 CHF
ORIGTA313824
Anti-EPB41L2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: EPB41L2 (erythrocyte membrane protein band 4.1-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR