Anti-DPYSL2 Rabbit Polyclonal Antibody
Supplier: OriGene
ORIGTA313766EA 642 CHF
ORIGTA313766
Anti-DPYSL2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: DPYSL2 (dihydropyrimidinase-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat