Anti-DNM1 Rabbit Polyclonal Antibody
Supplier: OriGene
ORIGTA313758EA 642 CHF
ORIGTA313758
Anti-DNM1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000
Type: Primary
Antigen: DNM1 (dynamin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat