Anti-BRCA2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313520EA 656 CHF
ORIGTA313520
Anti-BRCA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: BRCA2 (breast cancer 2, early onset)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR