Anti-NR3C1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313370EA 642 CHF
ORIGTA313370
Anti-NR3C1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 500-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: NR3C1 (nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR