Anti-ITPKC Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312655EA 656 CHF
ORIGTA312655
Anti-ITPKC Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ITPKC (inositol-trisphosphate 3-kinase C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR