Anti-BAX Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312599EA 619 CHF
ORIGTA312599
Anti-BAX Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR