Anti-ATXN7L1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312575EA 632 CHF
ORIGTA312575
Anti-ATXN7L1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: ATXN7L1 (ataxin 7-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR