Anti-FAM84B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312562EA 656 CHF
ORIGTA312562
Anti-FAM84B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: FAM84B (family with sequence similarity 84, member B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR