Anti-P2RY13 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312556EA 642 CHF
ORIGTA312556
Anti-P2RY13 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: P2RY13 (purinergic receptor P2Y, G-protein coupled, 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR