Anti-RASSF6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312555EA 632 CHF
ORIGTA312555
Anti-RASSF6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: RASSF6 (Ras association (RalGDS/AF-6) domain family member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR