Anti-CCNB1IP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312550EA 642 CHF
ORIGTA312550
Anti-CCNB1IP1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: CCNB1IP1 (cyclin B1 interacting protein 1, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR