Anti-PTK2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312487EA 656 CHF
ORIGTA312487
Anti-PTK2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: HC: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR