Anti-CTTN Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312466EA 619 CHF
ORIGTA312466
Anti-CTTN Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: CTTN (cortactin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR