Anti-RAD17 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312462EA 656 CHF
ORIGTA312462
Anti-RAD17 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: RAD17 (RAD17 homolog (S. pombe))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR