Anti-APOA5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312455EA 642 CHF
ORIGTA312455
Anti-APOA5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR