Anti-C2orf40 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312430EA 632 CHF
ORIGTA312430
Anti-C2orf40 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: C2orf40 (chromosome 2 open reading frame 40)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR