Anti-PPP1R1B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312427EA 642 CHF
ORIGTA312427
Anti-PPP1R1B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: PPP1R1B (protein phosphatase 1, regulatory (inhibitor) subunit 1B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR