Anti-XRCC4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312413EA 619 CHF
ORIGTA312413
Anti-XRCC4 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: XRCC4 (X-ray repair complementing defective repair in Chinese hamster cells 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR