Anti-RELA Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312411EA 642 CHF
ORIGTA312411
Anti-RELA Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR