Anti-RPS6KA2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312398EA 656 CHF
ORIGTA312398
Anti-RPS6KA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: RPS6KA2 (ribosomal protein S6 kinase, 90kDa, polypeptide 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR