Anti-OTUB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312383EA 642 CHF
ORIGTA312383
Anti-OTUB1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OTUB1 (OTU deubiquitinase, ubiquitin aldehyde binding 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR