Anti-CASP10 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312376EA 632 CHF
ORIGTA312376
Anti-CASP10 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:300-1:1000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: CASP10 (caspase 10, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR