Anti-MRPS27 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312373EA 642 CHF
ORIGTA312373
Anti-MRPS27 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: MRPS27 (mitochondrial ribosomal protein S27)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR