Anti-VWA5A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312370EA 642 CHF
ORIGTA312370
Anti-VWA5A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: VWA5A (von Willebrand factor A domain containing 5A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR