Anti-RPS6KA6 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312367EA 642 CHF
ORIGTA312367
Anti-RPS6KA6 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: RPS6KA6 (ribosomal protein S6 kinase, 90kDa, polypeptide 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR