Anti-AHSA1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312349EA 642 CHF
ORIGTA312349
Anti-AHSA1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: AHSA1 (AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR