Anti-HSP90AB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312344EA 656 CHF
ORIGTA312344
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunoprecipitation: Various DilutionImmunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: HSP90AB1 (heat shock protein 90kDa alpha (cytosolic), class B member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR