Anti-DNAJB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312301EA 656 CHF
ORIGTA312301
Anti-DNAJB1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:300-1:1000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: DNAJB1 (DnaJ (Hsp40) homolog, subfamily B, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR