Anti-MTNR1A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312291EA 642 CHF
ORIGTA312291
Anti-MTNR1A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: MTNR1A (melatonin receptor 1A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR