Anti-MAPT Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312283EA 656 CHF
ORIGTA312283
Anti-MAPT Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5100

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR