Anti-STAM2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312253EA 656 CHF
ORIGTA312253
Anti-STAM2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:1000

Type: Primary
Antigen: STAM2 (signal transducing adaptor molecule (SH3 domain and ITAM motif) 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR