Anti-MAGEA1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312178EA 656 CHF
ORIGTA312178
Anti-MAGEA1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: MAGEA1 (melanoma antigen family A, 1 (directs expression of antigen MZ2-E))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR