Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312138EA 642 CHF
ORIGTA312138
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR