Anti-TGFBR3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312087EA 656 CHF
ORIGTA312087
Anti-TGFBR3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: TGFBR3 (transforming growth factor, beta receptor III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR