Anti-TFAM Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312086EA 656 CHF
ORIGTA312086
Anti-TFAM Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: TFAM (transcription factor A, mitochondrial)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR